Anti-TBX5 Mouse Monoclonal Antibody [clone: 1G10]
Catalog # ABNOH00006910-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:T-box 5
- Antigen symbol:TBX5
- Clonality:Monoclonal
- Clone:1G10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- ChIP:Yes
- Gene ID:6910
- Antigen synonyms:TBX5|HOS
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TBX5
Clonality: Monoclonal
Clone: 1G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human