Anti-SUMO2 Mouse Polyclonal Antibody
ABNOH00006613-A01
: Abnova
- Antibody type:Primary
- Antigen name:Small ubiquitin-related modifier 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:6613
- Antigen synonyms:Sentrin-2|SUMO-2|HSMT3|SMT3 homolog 2|Small ubiquitin-related modifier 2|Ubiquitin-like protein SMT3B|SUMO-3|SMT3B|SMT3H2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: SUMO2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human