Anti-PSMB8 Mouse Monoclonal Antibody [clone: 1B3]
Catalog # ABNOH00005696-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Proteasome 20S Beta 8
- Antigen symbol:PSMB8
- Clonality:Monoclonal
- Clone:1B3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5696
- Antigen synonyms:ALDD|JMP|D6S216E|macropain) Subunit Beta Type 8|LMP7|PSMB5i|NKJO|Proteasome (prosome|D6S216|RING10
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: PSMB8
Clonality: Monoclonal
Clone: 1B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human