Order Entry
Portugal
ContactUsLinkComponent
Anti-IFNA2 Mouse Monoclonal Antibody [clone: 2B1]
Anti-IFNA2 Mouse Monoclonal Antibody [clone: 2B1]
Catalog # ABNOH00003440-M26
Supplier:  Abnova
undefined
Anti-IFNA2 Mouse Monoclonal Antibody [clone: 2B1]
Catalog # ABNOH00003440-M26
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interferon alpha 2
  • Antigen symbol:
    IFNA2
  • Clonality:
    Monoclonal
  • Clone:
    2B1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    3440
  • Antigen synonyms:
    IFN-alphaA|INFA2|IFNA2B|IFNA
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: IFNA2
Clonality: Monoclonal
Clone: 2B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human