Anti-HES1 Mouse Monoclonal Antibody [clone: 3A3]
Catalog # ABNOH00003280-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Hairy and enhancer of split 1
- Antigen symbol:HES1
- Clonality:Monoclonal
- Clone:3A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3280
- Antigen synonyms:HHL|Transcription factor HES-1|HL|HRY|Hairy homolog|Hairy and enhancer of split 1|Hairy-like
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: HES1
Clonality: Monoclonal
Clone: 3A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat