Order Entry
Portugal
ContactUsLinkComponent
Anti-FOXA2 Mouse Monoclonal Antibody [clone: 8H5]
Anti-FOXA2 Mouse Monoclonal Antibody [clone: 8H5]
Catalog # ABNOH00003170-M03A
Supplier:  Abnova
undefined
Anti-FOXA2 Mouse Monoclonal Antibody [clone: 8H5]
Catalog # ABNOH00003170-M03A
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Forkhead Box A2
  • Antigen symbol:
    FOXA2
  • Clonality:
    Monoclonal
  • Clone:
    8H5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    3170
  • Antigen synonyms:
    Forkhead box protein A2|Hepatocyte nuclear factor 3-beta|HNF3B|HNF-3-beta|TCF3B
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: FOXA2
Clonality: Monoclonal
Clone: 8H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human