Order Entry
Portugal
ContactUsLinkComponent
Büchner funnels, KIMAX®, KIMBLE®
Büchner funnels, KIMAX®, KIMBLE®
Catalog # SCERSP28400-601
Supplier:  DWK Life Sciences
Büchner funnels, KIMAX®, KIMBLE®
Catalog # SCERSP28400-601
Supplier:  DWK Life Sciences

Specifications

  • Plate Ø:
    40 mm
  • Porosity:
    40 - 60 µm
  • Capacity:
    60 ml
  • Pk:
    6

Specifications

About this item

KIMAX®, borosilicate glass.

  • With KIMFLOW® fritted disc
  • With heavy walled stems
  • Available with marking spot
  • Beaded rims for extra strength

This funnel is used wherever the filter paper and membranes are unable to withstand the chemical attack of the filtrate.