Order Entry
Portugal
ContactUsLinkComponent
Pocket pH/°C meter, HI 9214
Pocket pH/°C meter, HI 9214
Catalog # HANNHI9214
Supplier:  Hanna
Pocket pH/°C meter, HI 9214
Catalog # HANNHI9214
Supplier:  Hanna

Specifications

  • Temperature range:
    −5,0...+70 °C
  • Description:
    Pocket pH/mV tester, HI 9214, with electrode
  • Model:
    HI 9214
  • Temperature accuracy:
    ±0,5 °C
  • Temperature resolution:
    0,1 °C
  • Automatic buffer recognition:
    Yes
  • Display:
    LCD
  • Power supply:
    9 V alkaline battery for 700 hours of continuous use
  • Temperature compensation:
    Automatic
  • Weight:
    0,16 kg
  • W×D×H:
    185×42×26 mm
  • pH accuracy:
    ±0,02
  • pH calibration:
    Automatic, 1 or 2 points with 3 memorised buffer values (4,01 / 7,01 / 10,01)
  • pH range:
    −2,00...+16,00
  • pH resolution:
    0,01
  • Pk:
    1

Specifications

About this item

HI 9214 is a compact, stick-type, microprocessor-based pH/°C meter designed to be easily carried anywhere. The included HI 12170 pH-electrode has a built-in temperature sensor for quick and accurate temperature measurements, and automatic temperature compensation.

  • Interchangeable plastic-body electrode
  • Simple controls on the front panel
  • Low battery detector
  • No trimmers: Simple calibration procedure
  • Three memorised buffer values (pH 4,01, pH 7,01, pH 10,01) and automatic buffer recognition to avoid errors during calibration
  • Lightweight ABS plastic housing which withstands the effects of most chemicals in the lab and industry
Delivery: Supplied with HI 12170 pH electrode, soft carrying case and 9 V battery and instruction manual.