SMD tweezers, Erem®
Supplier: Basan
Certificates
About this item
Stainless steel. These special tweezers are ideal for handling chips or miniature SMD components. Suitable for mounting on SMD boards or ceramic substrates.
- With anti-glare finish, non magnetic
- With angled head and blunt edges to avoid board damage
Frequently Bought Together