Pump tubing, Tygon® E-1000
Tygon®, translucent.
- Soft and flexible
- Low-temperature resistant for low level cryogenic applications
- Stays flexible at temperatures as low as –55 °C
- Resistant to corrosive chemicals for laboratory applications
- Low durometer for use in low-torque pump applications
Tygon® E-1000 tubing is an excellent solution to applications requiring complicated laboratory set-ups with sharp radius curves and multiple directional changes. This tubing resists twisting and collapse, which are common problems when using other tubing products. This tubing delivers superior performance in a formulation that contains non-DEHP [Bis (2-ethylhexyl) phthalate] plasticisers.
Temperature resistance: –73 to +52 °C
Hardness (Shore durometer A): 40
Density: 1,10
: Meets FDA 21 CFR 175.300 requirements for food contact.
Frequently Bought Together