Order Entry
Portugal
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid 25,5 g/l in aqueous solution

L(+)-Ascorbic acid 25,5 g/l in aqueous solution

Glass bottle, 0,5 mL
Catalog # CABRCC-4398
Marketsource
Supplier:   Lonza
L(+)-Ascorbic acid 25,5 g/l in aqueous solution
Catalog # CABRCC-4398
Supplier:  Lonza
Marketsource
CAS Number:  50-81-7
L(+)-Ascorbic acid 25,5 g/l in aqueous solution
L(+)-Ascorbic acid 25,5 g/l in aqueous solution

Glass bottle, 0,5 mL

CAS Number:  50-81-7
Price
128,00 €
Log in for availability
1 * 0,5 mL
Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-066-2
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    0,5 mL

Specifications