Ethidium bromide 10 mg/ml in aqueous solution for molecular biology
Catalog # PROMH50410010
Supplier: Promega
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
Specifications
About this item
:
:394.31 g/mol
EINECS:214-984-6
:GHS08
CAS Number:1239-45-8
Formula:C₂₁H₂₀BrN₃
MDL Number:MFCD00011724