Order Entry
Portugal
ContactUsLinkComponent
Octyltin trichloride
Octyltin trichloride
Catalog # PROODEMS-03443-0.3
CAS Number:  
Octyltin trichloride
Catalog # PROODEMS-03443-0.3
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    300 mg
  • Seq:
    2625

Specifications

About this item

 :
EINECS:221-435-4
CAS Number:3091-25-6
Formula:C₈H₁₇Cl₃Sn