Order Entry
Portugal
ContactUsLinkComponent
VWR® Q Path® Stick on (Cell adhesive) for Microscopy
VWR® Q Path® Stick on (Cell adhesive) for Microscopy
Catalog # 11047600.
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Q Path® Stick on (Cell adhesive) for Microscopy
Catalog # 11047600.
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    Stick on (Cell adhesive) Q Path®
Specifications
About this item
Glue for improving adherance of histologicals sections to slides. Alternative to albumin-glycerol.