
Methyl-3,5-dibromobenzoate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:51329-15-8
:
Formula:C₈H₆Br₂O₂
MDL Number:MFCD00082648
:293.94 g/mol
:GHS07
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:61.0 to 65.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together