1,1'-Bis(diisopropylphosphino)ferrocene ≥98.0% (by titrimetric analysis)
: TCI
:97239-80-0
:
:C₂₂H₂₈FeP₂
:MFCD02684559
:410.25 g/mol
- Appearance:Orange to Amber to Dark red powder to crystal
- Purity (Chelometric Titration):min. 98.0 %
- Melting point:48.0 to 52.0 deg-C
Frequently Bought Together