Specifications
- Packed:Pack of 10 pcs.
- Capacity:-
- Colour:White
- Description:For Eppendorf Tubes® 5.0 mL
- Seq:05
- Environmentally Preferable:Low Manufacturing Impact
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Pk:10
- RCF max.:25000×g
Specifications
About this item
High quality polypropylene, transparent.
- Easier processing of samples up to 5,0 ml
- Higher yields in DNA isolation, especially with samples with limited quantity of source material
- Convenient and safe cell culture applications
- Easier sample access lowers the risk of contamination
- Preparation of mastermixes and buffers
- Less storage space needed
- Safe cell and tissue lysis
Eppendorf tubes are the benchmark for simple and safe sample preparation from 0,5 to 2,0 ml. For larger volumes, 15 and 50 ml conical tubes are available. With medium sample volumes you may face an issue; they need to be processed with large conical screw cap tubes - impractical, inconvenient and often prone to contamination. Eppendorf now offers the missing links; the Eppendorf Tubes® 5,0 ml. This product fills the gap between existing tube versions and enables the simple and safe processing of sample volumes up to 5,0 ml. Simple, practical and ergonomic single-hand operation. Large labelling area. Hinged lid for minimised sample evaporation during storage and incubation in a wide range of temperatures from −86 to +80 °C. Material is free from plasticisers, biocides or mould release agents, for reliable test results.