Order Entry
Portugal
Orders LinkContactUsLinkComponent
French square bottles, clear, WHEATON®
French square bottles, clear, WHEATON®
French square bottles, clear, WHEATON®
French square bottles, clear, WHEATON®
French square bottles are manufactured from clear soda lime glass that conforms to USP Type III requirements. Design allows for full utilisation of any storage area, maximising space efficiency. Each case of bottles is packaged in convenience packs with caps attached to maintain cleanliness. A partitioned carton protects bottles from scratches during shipping, and they become useful for storing samples. Each case of bottles also includes a hazardous communication label and tamper evident seal if labelling and/or sealing is required. Extra screw caps can be purchased separately.
 : Caps are supplied attached to bottles.
Product Family Options
Clear french square bottles with rubber lined black phenolic cap
  • Capacity
    Thread
    Ext.Ø
    Height
    Colour
  • 15 ml
    20-400
    26 mm
    62 mm
    Clear
    1 * 48 unid.
    1 * 48 unid. - 92,20 €
    92,20 €
    1 * 48 unid.
       92,20 €
  • 30 ml
    24-400
    31 mm
    72 mm
    Clear
    1 * 48 unid.
    1 * 48 unid. - 93,20 €
    93,20 €
    1 * 48 unid.
       93,20 €
  • 60 ml
    28-400
    39 mm
    87 mm
    Clear
    1 * 48 unid.
    1 * 48 unid. - 108,00 €
    108,00 €
    1 * 48 unid.
       108,00 €