Order Entry
Portugal
ContactUsLinkComponent
Bottles for Canada balsam
Bottles for Canada balsam
Catalog # KAVA632425006100
Supplier:  Kavalier
Bottles for Canada balsam
Catalog # KAVA632425006100
Supplier:  Kavalier

Specifications

  • Ext.Ø:
    63 mm
  • Capacity:
    100 ml
  • Height:
    115 mm
  • Pk:
    1

Specifications

About this item

Glass, clear.