Chlorate standard solution, 1,000 mg/L ClO₃- in Water
Catalog # CMPLF750.1K.W.5
Supplier: CUSTOM MADE CHEMICALS LAB
CAS Number:
Specifications
- Concentration:1000 mg/l
- Matrix:H₂O
- Pack type:Glass ampoule
- Pk:1 Ampoul
- Description:Chlorate standard solution (ClO₃⁻)