Buffer, TRIS-glycine-SDS solution 10X concentrate (TGS buffer)
TRIS-glycine-SDS buffer solution 10X concentrate (TGS buffer), 4 L
Catalog # CABR50882
Marketsource
Supplier: Lonza
Buffer, TRIS-glycine-SDS solution 10X concentrate (TGS buffer)
Catalog # CABR50882
Supplier: Lonza
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:4 L
- Description:TRIS-glycine-SDS buffer solution 10X concentrate (TGS buffer)



