
1,4,8,11-Tetraazacyclopentadecane ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:15439-16-4
:
Formula:C₁₁H₂₆N₄
MDL Number:MFCD00010049
:214.35 g/mol
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Nonaqueous Titration):min. 97.0 %
- Melting point:99.0 to 103.0 deg-C
Specification Test Results
Related Information
Frequently Bought Together