Order Entry
Portugal
Orders LinkContactUsLinkComponent
Dapsone ≥98.0% (by HPLC, titration analysis)

Dapsone ≥98.0% (by HPLC, titration analysis)

Irritant
Health
Environment
Supplier:   TCI
Dapsone ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
Dapsone ≥98.0% (by HPLC, titration analysis)
Dapsone ≥98.0% (by HPLC, titration analysis)

3 Options Available Below

Product Details & Documents
Synonyms:  4,4'-sulphonyldianiline, 4,4'-Diamino diphenyl sulphone, 4-[(4-Aminophenyl)sulphonyl]aniline, DDS, 4,4'-Sulphonyldianiline, 1-Amino-4-((4-aminophenyl)sulphonyl)benzene, 4-Aminophenyl sulphone
  • CAS Number:80-08-0
  • Formula:C₁₂H₁₂N₂O₂S
  • MDL Number:MFCD00007887
  • Molecular Weight:248.31 g/mol
  • Merck Index:14,02822
  • EINECS:201-248-4
  • UN:3077
  • ADR:9,III
Specifications for Product Family

Specification Test Results

  • Appearance:
    White to Light yellow powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    173.0 to 181.0 deg-C
  • Solubility in hot Methanol:
    almost transparency
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 20,00 €
    20,00 €
    1 * 25 g
       20,00 €

  • 100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 38,00 €
    38,00 €
    1 * 100 g
       38,00 €

  • Glass bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 116,00 €
    116,00 €
    1 * 500 g
       116,00 €