Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')europium(III) ≥95.0% (by titrimetric analysis) NMR shift reagent
: TCI
:15522-71-1
:
:C₃₃H₅₇EuO₆
:MFCD00000019
:701.766 g/mol
:239-565-5
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (Chelometric Titration):min. 95.0 %
- Melting point:188.0 to 192.0 deg-C
Frequently Bought Together