Order Entry
Portugal
Orders LinkContactUsLinkComponent
wells-industrial-gloves-hero.jpg
Industrial Gloves
Protect your hands in many different applications with a large selection of gloves and sleeves.
Wells Lamont Industrial Gloves | Avantor
Wells Lamont products can help protect your hands in many industrial situations. Options include cut resistant, leather, mechanics, impact, heat resistant, palm dipped, and general purpose. We not only support critical environment applications but also maintenance, food processing, manufacturing, metal handling, welding, metal stamping, shipping, oil and gas, and many more.
Leather-Glove.jpg
Leather Glove
Our Y0122 is a high quality grain cowhide drivers glove that offers high abrasion and puncture resistance.
Metal--Mesh--Glove.jpg
This glove offers superior cut resistance and features a patented self tensioning cuff that is easier to put on and more hygenic.
Heat-glove.jpg
The terry cloth 636HR offers excellent heat resistance up to 350F and features a 5" gauntlet for protecting the wrist area.
Palm-Dipped-Glove.jpg
The Y9265 has a polyurethane palm for grip and offers ANSI A2 cut resistance.
SleeveANSI.jpg
The Armguard II Medium Duty is a 20" high level cut resistant sleeve that is made in the USA.