Order Entry
Portugal
Orders LinkContactUsLinkComponent
techsolv-AD1.jpg
TexSolv & CoreSafe Chemical Protection
SW® is advancing materials science and new manufacturing processes to develop chemical protection products with exact safety specifications while maintaining a focus on work comfort and hand health. Look for our chemical offering to expand in the near future.
NewTrile & CoreSafe Chemical Protection | Avantor
TexSolv® nitrile provides broad spectrum chemical protection and tip to cuff consistency. CoreSafe® features easy-to-use natural rubber protection.
Product selection
EcoTek-Sustainable-Technology.jpg
Chemical-resistant unlined nitrile gloves. Gloves are 11mil thick and double chlorinated with recessed diamond grip and a straight cuff.
texsol.jpg
Chemical-resistant unlined nitrile gloves. Gloves are 15mil thick and double chlorinated with recessed diamond grip and a straight cuff.
texsol-f15.jpg
Flock-lined chemical-resistant nitrile gloves. Gloves are 15mil thick and double chlorinated with recessed diamond grip and a straight cuff.
CoreSafe-F13Y.jpg
Yellow flock-lined chemical-resistant latex gloves are 13mil thick with a fish scale grip pattern and double chlorinated with a straight cuff.
CoreSafe-F28O.jpg
Orange flock-lined chemical-resistant latex gloves are 28mil thick with a fish scale grip pattern and double chlorinated with a straight cuff.
CoreSafe-U17N.jpg
Chemical-resistant latex gloves with natural coloring are 17mil thick and double chlorinated with a fish scale grip pattern.
CoreSafeF28YB.jpg
Flock-lined blue neoprene over yellow latex chemical-resistant gloves are 28mil with a raised diamond grip pattern and silicone-free.