Order Entry
Portugal
Orders LinkContactUsLinkComponent
Bucket-Liners.jpg
Liners for Contamination Control
Micronova Bags and Liners are used for a variety of containment functions in the cleanroom and aseptic areas. The Liners, available sterile, are used for storage of raw materials, disinfectant containment, disposal, and segregation of biohazardous materials.
Liners for Contamination Control | Avantor
BucketLiner_in_Yellow_B7_Micronova_200.jpg
BucketLiners™
An impermeable barrier made from LDPE/Nylon laminate is ideal for lining buckets containing liquid solutions or to line totes for transporting product parts or raw materials. Excellent for liquid containment and material transport.
BinLiner_inside_Stand_Micronova_200_1.jpg
BinLiner™ Cleanroom Bags
These LDPE liners are ideal for a variety of uses ranging from the collection and disposal of waste to the transport of parts or raw materials in cleanrooms and aseptic environments. Available gamma irradiated or non-irradiated in multiple thicknesses and capacities.
Biohazard_Red_Bag_Micronova_200.jpg
BioHazard and Hazard BinLiner™ Bags
These LDPE liners are ideal for a variety of uses ranging from the collection and disposal of waste to the transport of parts or raw materials in cleanrooms and aseptic environments. Available gamma irradiated or non-irriadated in multiple thicknesses and capacities.
BinLiner_inside_Stand_Micronova_200.jpg
Stainless Steel BinLiner™ Stands
Bags, with easy-to-clean plastic corners and stainless steel supports that do not touch the floor. ideal for cleanroom material disposal, garment collection, and raw material or product storage. Multiple sizes available.