Order Entry
Portugal
Orders LinkContactUsLinkComponent
Tapes.jpg
Cleanroom Tapes for Every Type of Application
Micronova's range of cleanroom pressure sensitive and barrier tapes include low residue and low outgassing tapes for production, marking, sealing, and a number of other applications. The tapes are wound on a plastic core to help eliminate particles in a controlled environment, and may be gamma irradiated for aseptic environments.
Cleanroom Tapes for Every Type of Application | Avantor
Vinyle.jpg
Cleanroom Vinyl Tape
Ultra processed and designed to limit particulate and bio-burden counts in cleanrooms and clean environments. Ideal for batching, color coding, packaging, etc. Matte finish can be written on with cleanroom pen and it has a clean peel from most surfaces.
Sleeve-Seal.jpg
Irradiated M-Zone™ CuffStrip™ Tape
Pre-cut, 18-inch, conformable LDPE tape strips are used to ensure an effective seal between the glove and garment in critical environments and quick release without damaging garments or gloves. Low-tack acrylic adhesive for easy garment release, gamma irradiated.
MicroSeal.jpg
MicroSeal™ Perforated Cuff and Plate Sealing Tape
Perforated at 7.6 cm (3 inch) intervals, this conformable tape tears at the optimum length to secure glove to garment or to seal plates. Low-density, low-outgassing tape. Available gamma irradiated for aseptic environments. Suitable for pharmaceutical production.
Autoclave-Indicator.jpg
Lead-Free Autoclave Indicator Tape
A proven method of positively confirming completion of the sterilization cycle in both gravity discharge and vacuum autoclaves. Lead-free, saturated crepe paper with synthetic polyisoprene adhesive it has green indicator lines that change to black when sterilization is achieved Certifications: RoHS Compliant.
Cleanroom-Tape.jpg
Cleanroom Polyethylene Tape
Low density polyethylene tape is low in halogens and leachable chlorides. Special construction lessens the chance of outgassing. This pressure sensitive tape has good chemical resistance and conformability.
rradiated-Cleanroom.jpg
Irradiated Cleanroom Vinyl Tape
Ultra processed and designed to limit particulate and bio-burden counts in cleanrooms and clean environments. Gamma irradiated for aseptic environments, it can be used for marking and labeling. Matte finish can be written on with cleanroom pen and it has a clean peel from most surfaces.
Extreme-Temperature.jpg
Extreme Temperature Polyimide Cleanroom Tape
Designed applications: -100°F to 500°F (-70°C to 260°C). Ideal for heat-sealing, solder mask for circuit boards, flame-resistant electrical insulation, lamination, cold storage, lyophilization, and freeze drying.
NovaStat.jpg
NovaStat™ Antistatic Tape
Specially designed for sealing and packaging in ESD-sensitive environments, with non-hygroscopic properties, cellophane backing and a rubber based adhesive.
CGT-Conductive.jpg
CGT™ Conductive Grid Tape
Heavy duty, antistatic tape with conductive grids embedded between two antistatic layers, for use on sensitive devices in ESD-sensitive environments. Protects sensitive devices that would be otherwise damaged by normal insulating tape.