Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
INITIAL LOAD
Catalog # 78046-753
Supplier:
Bio X Cell
New Product
Expand image
INITIAL LOAD
Catalog # 78046-753
Supplier:
Bio X Cell
Supplier Number:
BP0366-R050MG
New Product