$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
INITIAL LOAD
78046-716
:
Bio X Cell
New Product
INITIAL LOAD
78046-716
:
Bio X Cell
:
BP0003-3-R005MG
New Product