Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Sodium hydroxide ≥99%

Sodium hydroxide ≥99%

Plastic bottle, 10 kg
Catalog # CH00271-10KG
New Product
Supplier #:  00271-10KG
Sodium hydroxide ≥99%
Catalog # CH00271-10KG
New Product
CAS Number:  1310-73-2
Sodium hydroxide ≥99%
Sodium hydroxide ≥99%

Plastic bottle, 10 kg

CAS Number:  1310-73-2
Price
$161.76
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Sodium hydroxide, commonly known as caustic soda or lye, is a highly versatile and essential chemical compound widely used across various industries. This strong alkaline substance is known for its ability to effectively neutralize acids, making it invaluable in chemical manufacturing processes, such as the production of soaps, detergents, and various organic chemicals. Its high solubility in water and exothermic reaction upon dissolution enhance its effectiveness in applications like pH regulation, where it is employed to maintain optimal conditions in water treatment facilities and industrial processes.

In addition to its role in manufacturing, sodium hydroxide is crucial in the food industry for processes such as food processing and the production of certain food additives. Its ability to act as a powerful cleaning agent also makes it a popular choice for industrial cleaning applications. Researchers and professionals appreciate sodium hydroxide for its reliability and efficiency, particularly in laboratory settings where it is used for titrations and other analytical procedures. With its broad range of applications and effectiveness, sodium hydroxide remains a staple in both industrial and research environments.

Store at RT.

Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Storage Temperature:Room Temperature
  • UN:1823
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    10 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.