Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Boc-hydrazide ≥99%

Plastic bottle, 10 kg
Catalog # CH02286-10KG
New Product
 :  02286-10KG
Boc-hydrazide ≥99%
Catalog # CH02286-10KG
New Product
CAS Number:  870-46-2
Boc-hydrazide ≥99%
Boc-hydrazide ≥99%

Plastic bottle, 10 kg

CAS Number:  870-46-2
Price
$3,664.88
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Boc-hydrazide is a versatile chemical compound widely utilized in organic synthesis and pharmaceutical research. Its unique structure, featuring a tert-butyl group, enhances its stability and solubility, making it an ideal candidate for various applications. This compound is particularly valuable in the synthesis of hydrazones and other nitrogen-containing heterocycles, which are essential in the development of agrochemicals and pharmaceuticals. Researchers appreciate Boc-hydrazide for its ability to facilitate the formation of peptide bonds, thereby playing a crucial role in peptide synthesis and drug development.

Additionally, Boc-hydrazide serves as a protecting group in organic reactions, allowing for selective functionalization of other reactive sites. Its ease of removal under mild conditions makes it a preferred choice among chemists. With its broad applicability in medicinal chemistry, including the design of inhibitors and other bioactive compounds, Boc-hydrazide stands out as a key reagent in the toolkit of researchers and industry professionals alike.

Store at 0 to 8 °C.

Synonyms:  tert-Butyl hydrazinecarboxylate
  • CAS Number:870-46-2
  • Formula:C₅H₁₂N₂O₂
  • MDL Number:MFCD00007593
  • Molecular Weight:132.2 g/mol
  • Storage Temperature:Refrigerator
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    10 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.