Uridine ≥99% (by UV method)
Product Details & Documents
Uridine is a naturally occurring nucleoside that plays a crucial role in various biological processes, including RNA synthesis and cellular metabolism. This compound is recognized for its potential applications in neuroprotection and cognitive enhancement, making it a valuable asset in both pharmaceutical research and dietary supplement formulations. Uridine is known to support brain health by promoting the synthesis of phospholipids, which are essential for neuronal membrane integrity and function. Additionally, it has been studied for its potential benefits in conditions such as depression and cognitive decline, providing a promising avenue for therapeutic development.
Researchers and industry professionals can leverage Uridine's unique properties in a variety of applications, from developing innovative nootropic supplements to exploring its role in regenerative medicine. Its ability to enhance synaptic plasticity and support mitochondrial function positions Uridine as a compound of interest in the fields of neuroscience and metabolic health. With its growing recognition in the scientific community, Uridine offers exciting possibilities for advancing health and wellness solutions.
Store at room temperature.
- CAS Number:58-96-8
- Formula:C₉H₁₂N₂O₆
- MDL Number:MFCD00006526
- Molecular Weight:244.2 g/mol
- Storage Temperature:Room Temperature
Specifications
Specifications
- Pack type:Plastic bottle
- Size:100 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



