CELL LINE 93T449 HUMAN FIBROBLAST LIKE
Catalog # MSPP-CRL-3043
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
CELL LINE 93T449 HUMAN FIBROBLAST LIKE
Catalog # MSPP-CRL-3043
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
Supplier Number: CRL-3043
Frequently Bought Together
BUFFER MES SDS RUNNING 20X SOLUTION 1L
Supplier: G-Biosciences
MARKER MAURICE CIEF PI-8.40 LYOPH 210UL
Supplier: PROTEIN SIMPLE MS
2D Electrophoresis Reagents, G-Biosciences
Supplier: G-Biosciences
RNA Pre-Wash Buffer, Zymo Research
Supplier: Zymo Research
Electrophoresis Running Buffers, G-Biosciences
Supplier: G-Biosciences
RBC Lysis Buffer, Zymo Research
Supplier: Zymo Research
Tris Buffers, Sterile
Supplier: G-Biosciences
IPG Buffers, Cytiva
Supplier: Cytiva
Zyppy Elution Buffer, Zymo Research
Supplier: Zymo Research
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
GelBond® Film
Supplier: Lonza
OLIGO BINDING BUFFER (40 ML)
Supplier: Zymo Research
KIT POWER SNAP ELECTROPHORSIS EX1% 240V
Supplier: Invitrogen
KIT DENSITY MARKER BEADS 6 VIALS POLYETH
Supplier: COSPHERIC MS