Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Nitinol, tube Int.Ø: 0.6 mm, Ext.Ø: 0.8 mm, 0.1 mm

Nitinol, tube Int.Ø: 0.6 mm, Ext.Ø: 0.8 mm, 0.1 mm

Lenght 200 mm
Catalog # 78026-462
New Product
Supplier:   Goodfellow
Supplier #:  1000160829
Nitinol, tube Int.Ø: 0.6 mm, Ext.Ø: 0.8 mm, 0.1 mm
Catalog # 78026-462
Supplier:  Goodfellow
New Product
CAS Number:  
Nitinol, tube Int.Ø: 0.6 mm, Ext.Ø: 0.8 mm, 0.1 mm
Nitinol, tube Int.Ø: 0.6 mm, Ext.Ø: 0.8 mm, 0.1 mm

Lenght 200 mm

CAS Number:  
Price
$462.00
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Nitinol Tube is a tube product made of the shape memory alloy nitinol. This nickel-titanium alloy exhibits superelasticity and the ability to return to a predetermined shape when heated. Nitinol tubes from Goodfellow provide unique properties like biocompatibility, corrosion resistance, and fatigue resistance. These tubes find applications in the biomedical industry, robotics and aerospace where their ability to flex without yielding is advantageous. Goodfellow offers nitinol tubes ideal for prototyping and research and development projects.

  • Formula:Ni 55/Ti 45
Specifications

Specifications

  • Size:
    Lenght 200 mm

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.