Biuret ≥98%
Plastic bottle, 25 g
Catalog # CH22844-25G
New Product
Supplier: Chem-Impex International
: 22844-25G
Jump to:
Product Details & Documents
Synonyms: Carbamylurea, Allophanic acid amide
- CAS Number:108-19-0
- Formula:C₂H₅N₃O₂
- MDL Number:MFCD00007946
- Molecular Weight:103.08 g/mol
- Melting Pt:182...197 °C
- Storage Temperature:Ambient
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Appearance:White Crystalline Powder
- Infrared Spectrum:Conforms to Structure
- Loss on Drying:≤2.5%
- Assay:≥98%
- Purity by TLC:≥99%
Specifications
- Pack type:Plastic bottle
- Size:25 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



