CELL CRL-2322 EPITHELIAL FROZEN
MSPP-HCC1187
Frequently Bought Together
Mix & Go!™ E. coli Transformation Kit and Buffer Set
: Zymo Research
Buffer P3 (Yellow), Zymo Research
: Zymo Research
Color Prestained Protein Standard, Broad Range (10-250 kDa), New England Biolabs
: New England Biolabs (NEB)
ProSieve™ Protein Markers
: Lonza
Opal Prestained Protein Standard 10 to 245 kDa, Rockland Immunochemicals
: Rockland Immunochemical
DNA WASH BUFFER 6 ML
: Zymo Research
DNA Wash Buffer, Concentrate, Zymo Research
: Zymo Research
RIPA Lysis Buffer, Rockland Immunochemicals
: Rockland Immunochemical
Unstained Protein Standard, Broad Range, New England Biolabs
: New England Biolabs (NEB)
TrueBlot® IP/Western Blot, Rockland Immunochemicals
: Rockland Immunochemical
Bis-Tris Mini Protein Gels, GenScript®
: Genscript
Proteomelab™ SDS-MW Analysis Kit, SCIEX
: AB SCIEX LLC MS