Lithium chromate ≥99% (by assay)
Product Details & Documents
Lithium chromate is a vital compound used as a corrosion inhibitor in coatings and paints, enhancing durability in aerospace and automotive industries. It also serves as a reagent in analytical chemistry and contributes to advancements in battery technology. Ideal for professionals seeking reliable solutions in material enhancement.
Caution: Store at RT.
- CAS Number:14307-35-8
- Formula:Li₂CrO₄
- MDL Number:MFCD00016175
- Molecular Weight:129.87 g/mol
- Storage Temperature:Ambient
- UN:1479
Specifications
Specification Test Results
- Purity:≥99% (Assay)
- Apperance:Yellow Crystalline Powder
Specifications
- Pack type:Plastic bottle
- Size:5 kg
- Cat. No.:CH26487-5KG
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



