Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Uranium Standards, 100 µg/ml

Uranium, 100 µg/ml, 125 ml, 1% HNO₃, LDPE bottle
  77999-348
New Product
 :  MSU-100PPM-125ML
Uranium Standards, 100 µg/ml
  77999-348
New Product
 :  
Uranium Standards, 100 µg/ml
Uranium Standards, 100 µg/ml

Uranium, 100 µg/ml, 125 ml, 1% HNO₃, LDPE bottle

 :  
 
$57.00
   
Each(1/Itm/Bx)
 
   
 

 : ISO 17025/17034.

 

 

  • Concentration:
    100 µg/ml
  • Matrix:
    1% HNO₃
  • Pack type:
    LDPE bottle
  • Size:
    125 ml
  • Element:
    Uranium

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.