Orders LinkContactUsLinkComponent
Iron Standards
Iron Standards
Catalog # 77998-645
New Product
Supplier:   Inorganic Ventures
Supplier #:  CGFE10-125ML
Iron Standards
Catalog # 77998-645
Supplier:  Inorganic Ventures
New Product
CAS Number:  
Iron Standards
Iron Standards
CAS Number:  
Price
$104.00
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Certified reference material for Iron, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).
Certifications: ISO 17025/17034.