Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Erbium Standards

Erbium Standards

Erbium, 10,000 µg/ml, 30 ml, 7% HNO₃, LDPE bottle
Catalog # 77998-634
New Product
Supplier:   Inorganic Ventures
Supplier #:  CGER10-30ML
Erbium Standards
Catalog # 77998-634
Supplier:  Inorganic Ventures
New Product
CAS Number:  
Erbium Standards
Erbium Standards

Erbium, 10,000 µg/ml, 30 ml, 7% HNO₃, LDPE bottle

CAS Number:  
Price
$54.00
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Certified reference material for Erbium, traceable to NIST.

One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).

 : ISO 17025/17034.

Specifications

Specifications

  • Concentration:
    10,000 µg/ml
  • Matrix:
    7% HNO₃
  • Pack type:
    LDPE bottle
  • Size:
    30 ml
  • Element:
    Erbium

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.