Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Dysprosium Standards

Dysprosium Standards

Dysprosium, 1000 µg/ml, 125 ml, 7% HNO₃, LDPE bottle
Catalog # 77998-630
New Product
Supplier:   Inorganic Ventures
Supplier #:  CGDY1-125ML
Dysprosium Standards
Catalog # 77998-630
Supplier:  Inorganic Ventures
New Product
CAS Number:  
Dysprosium Standards
Dysprosium Standards

Dysprosium, 1000 µg/ml, 125 ml, 7% HNO₃, LDPE bottle

CAS Number:  
Price
$57.00
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Certified reference material for Dysprosium, traceable to NIST.

One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).

Certifications: ISO 17025/17034.

Specifications

Specifications

  • Concentration:
    1000 µg/ml
  • Matrix:
    7% HNO₃
  • Pack type:
    LDPE bottle
  • Size:
    125 ml
  • Element:
    Dysprosium

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.