Orders LinkContactUsLinkComponent
Dysprosium Standards
Dysprosium Standards
Catalog # 77998-629
New Product
Supplier:   Inorganic Ventures
Supplier #:  CGDY10-500ML
Dysprosium Standards
Catalog # 77998-629
Supplier:  Inorganic Ventures
New Product
CAS Number:  
Dysprosium Standards
Dysprosium Standards
CAS Number:  
Price
$325.00
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Certified reference material for Dysprosium, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).
Certifications: ISO 17025/17034.