Orders LinkContactUsLinkComponent
Dysprosium Standards
Catalog # 77998-627
New Product
Supplier:   Inorganic Ventures
 :  CGDY10-125ML
Dysprosium Standards
Catalog # 77998-627
Supplier:  Inorganic Ventures
New Product
CAS Number:  
Dysprosium Standards
Dysprosium Standards
CAS Number:  
Price
$110.00
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Certified reference material for Dysprosium, traceable to NIST.
One year expiration date (defined by supplier as the amount of time that it should remain in use after opening).
 : ISO 17025/17034.