BalanCD™ CHO Perfusion
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
BalanCD CHO Perfusion is a chemically defined medium that is optimized for the growth and production of suspension CHO cell cultures in continuous, steady-state, and N-1 perfusion cultures. As a high performing medium, it can achieve key perfusion metrics including cell-specific productivity, volumetric productivity, and cell-specific perfusion rate (CSPR). BalanCD CHO Perfusion medium meets the requirements of large-scale industrial cell cultures and is intended for long-term, continuous culturing of CHO cells. Support the growth and productivity of CHO cells in perfusion culture systems with BalanCD CHO Perfusion, a chemically defined, complete, and ready-to-use medium. Optimize and achieve key perfusion metrics including cell-specific productivity, volumetric productivity, and cell-specific perfusion rate (CSPR) for continuous, steady-state, and N-1 perfusion cultures.
Optimize perfusion processes to achieve cost-effective production of biotherapeutics
Complete, ready-to-use medium that supports cell growth, cell expansion, and production of monoclonal antibodies and recombinant proteins
Scalable from small shake flasks to large-scale perfusion bioreactors
Supports a variety of CHO subtypes
Delivers consistent, optimal cell productivities at 1 vessel volume per day (VVD)
Adaptable and scalable for a variety of perfusion culture platforms, including perfusion-mimic and perfusion-capable bioreactors
Available in liquid and powder formats
BalanCD CHO Perfusion is a chemically defined and animal component-free complete growth medium for Chinese Hamster Ovary (CHO) cells specifically designed for growth and productivity in a continuous steady-state perfusion process. BalanCD CHO Perfusion is provided without L-Glutamine to extend shelf life. The recommended L-Glutamine concentration to add is 4 to 8 mM (20 to 40 ml/L). Glucose should be monitored during culture as additional supplementation may be required. BalanCD CHO Perfusion has been designed to support high cell densities of CHO cell lines in perfusion-capable bioreactors at a minimum of 1 vessel volume per day. The medium does not contain antibiotics, antimycotics, protein hydrolysates or any other undefined components.
Packaging: Store at 2 to 8 °C. Shelf life: 12 months (liquid) or 24 months (powder) from date of manufacture.
Specifications
Specifications
- Media Format:Liquid
- Size:1 L
- Cat. No.:77975-346
- L-Glutamine:No
- Calcium:No
- HEPES:No
- Sodium pyruvate:No
- Glucose level:Without
- Sodium bicarbonate:No
- Magnesium:No
- Stable glutamine:No
- Media Base:Proprietary Mixture
- Phenol red:No
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



