Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Solution Bottles, Sterile

Solution Bottles, Sterile

Solution bottle, sterile, 1000 ml, Polystyrene, Clear
Catalog # 77973-188
New Product
Supplier #:  229785
Solution Bottles, Sterile
Catalog # 77973-188
New Product
Solution Bottles, Sterile
Solution Bottles, Sterile

Solution bottle, sterile, 1000 ml, Polystyrene, Clear

Price
$256.26
Log in for availability
Case of 24
Product Details & Documents

Solution bottles are key for storage, preparation, and dispensing of chemical solutions and reagents.

  • Compatible with bottle top filters or for use as solution storage containers
    Solution bottle neck size is 45 mm
    Curved bottle design for easy handling
Specifications
  • Capacity:
    1000 ml
  • Color:
    Clear
  • Description:
    Solution bottle, sterile
  • Material:
    Polystyrene
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.