Solution Bottles, Sterile
Solution bottle, sterile, 250 ml, Polystyrene, Clear
Catalog # 77973-184
New Product
Supplier: CELLTREAT Scientific Products
Supplier #: 229782
Jump to:
Product Details & Documents
Solution bottles are key for storage, preparation, and dispensing of chemical solutions and reagents.
- Compatible with bottle top filters or for use as solution storage containers
Solution bottle neck size is 45 mm
Curved bottle design for easy handling
Specifications
- Capacity:250 ml
- Color:Clear
- Description:Solution bottle, sterile
- Material:Polystyrene
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



