$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
RECOMBINANT HUMAN AMPK AL AB271362-10UG
ABCA_AB271362-10UG
:
Abcam
New Product
RECOMBINANT HUMAN AMPK AL AB271362-10UG
ABCA_AB271362-10UG
:
Abcam
:
AB271362-10UG
New Product