$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
RECOMBINANT HUMAN AMPK ALP AB268341-5UG
ABCA_AB268341-5UG
:
Abcam
New Product
RECOMBINANT HUMAN AMPK ALP AB268341-5UG
ABCA_AB268341-5UG
:
Abcam
:
AB268341-5UG
New Product