Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RECOMBINANT HUMAN AMPK ALP AB268334-5UG
ABCA_AB268334-5UG
:
Abcam
New Product
RECOMBINANT HUMAN AMPK ALP AB268334-5UG
ABCA_AB268334-5UG
:
Abcam
:
AB268334-5UG
New Product